Assay Details
Gene Symbol: LOC4345311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792678.1 | XP_015648164.1 | 2 - 3 | 699 | 84 | |
GenBank mRNA | AK068483.1 | - | 2 - 3 | 705 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792678.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
