Assay Details
Gene Symbol: LOC4345558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794730.2 | XP_015650216.1 | 1 - 2 | 99 | 119 | |
GenBank mRNA | AK059631.1 | - | 1 - 2 | 101 | 119 | |
AK099367.1 | - | 1 - 2 | 105 | 119 | ||
AK104578.1 | - | 1 - 2 | 99 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794730.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
