Assay Details
Gene Symbol: LOC4345558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794730.2 | XP_015650216.1 | 2 - 3 | 215 | 109 | |
GenBank mRNA | AJ575234.1 | - | 1 - 2 | 89 | 109 | |
AK059631.1 | - | 2 - 3 | 217 | 109 | ||
AK099367.1 | - | 2 - 3 | 221 | 109 | ||
AK104578.1 | - | 2 - 3 | 215 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
