Assay Details
Gene Symbol: LOC4345558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794730.2 | XP_015650216.1 | 3 - 4 | 335 | 96 | |
GenBank mRNA | AJ575234.1 | - | 2 - 3 | 209 | 96 | |
AK059631.1 | - | 3 - 4 | 337 | 96 | ||
AK099367.1 | - | 3 - 4 | 341 | 96 | ||
AK104578.1 | - | 3 - 4 | 335 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
