Assay Details
Gene Symbol: LOC4345558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794730.2 | XP_015650216.1 | 5 - 6 | 439 | 80 | |
GenBank mRNA | AJ575234.1 | - | 4 - 5 | 313 | 80 | |
AK099367.1 | - | 5 - 6 | 445 | 80 | ||
AK104578.1 | - | 5 - 6 | 439 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
