Assay Details
Gene Symbol: LOC4345558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794730.2 | XP_015650216.1 | 7 - 8 | 604 | 152 | |
GenBank mRNA | AJ575234.1 | - | 6 - 7 | 478 | 152 | |
AK059631.1 | - | 7 - 8 | 606 | 152 | ||
AK099367.1 | - | 7 - 8 | 610 | 152 | ||
AK104578.1 | - | 7 - 8 | 604 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
