Assay Details
Gene Symbol: LOC4345558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794730.2 | XP_015650216.1 | 8 - 9 | 726 | 92 | |
GenBank mRNA | AK059631.1 | - | 8 - 9 | 728 | 92 | |
AK099367.1 | - | 8 - 9 | 732 | 92 | ||
AK104578.1 | - | 8 - 9 | 726 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
