Assay Details
| Gene Symbol: LOC4324066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759775.2 | XP_015615261.1 | 3 - 4 | 1426 | 79 | |
| GenBank mRNA | AK069905.1 | - | 3 - 4 | 1433 | 79 | |
More Information
Set Membership: |
|

LOC4324066
Genomic Map