Assay Details
Gene Symbol: LOC4324675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761822.1 | XP_015617308.1 | 1 - 2 | 171 | 140 | |
GenBank mRNA | AK243597.1 | - | 1 - 2 | 123 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761822.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
