Assay Details
Gene Symbol: LOC4324675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761822.1 | XP_015617308.1 | 2 - 3 | 246 | 111 | |
GenBank mRNA | AK243597.1 | - | 2 - 3 | 198 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
