Assay Details
| Gene Symbol: LOC4324149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766432.2 | XP_015621918.1 | 11 - 12 | 3416 | 91 | |
| GenBank mRNA | AK064463.1 | - | 2 - 3 | 233 | 91 | |
More Information
Set Membership: |
|

LOC4324149
Genomic Map