Assay Details
| Gene Symbol: LOC4324149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766432.2 | XP_015621918.1 | 14 - 15 | 3612 | 61 | |
| GenBank mRNA | AK064463.1 | - | 4 - 5 | 916 | 61 | |
More Information
Set Membership: |
|

LOC4324149
Genomic Map