Assay Details
Gene Symbol: LOC4324001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766627.1 | XP_015622113.1 | 6 - 7 | 1689 | 123 | |
GenBank mRNA | AB154521.1 | - | 6 - 7 | 1631 | 123 | |
AK061744.1 | - | 4 - 5 | 899 | 123 | ||
AK067444.1 | - | 6 - 7 | 1695 | 123 | ||
AY578161.1 | - | 6 - 7 | 1637 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
