Assay Details
| Gene Symbol: LOC107278964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757024.1 | XP_015612510.1 | 1 - 2 | 204 | 68 | |
| GenBank mRNA | AK241684.1 | - | 1 - 2 | 155 | 68 | |
| Gene Symbol: LOC112935997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757060.1 | XP_015612546.1 | 1 - 2 | 198 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757024.1 transcripts.
Set Membership: |
|

LOC107278964
Genomic Map