Assay Details
| Gene Symbol: LOC4346094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793009.2 | XP_015648495.1 | 1 | 1983 | 116 | |
| GenBank mRNA | AK100965.1 | - | 1 | 1648 | 116 | |
More Information
Set Membership: |
|

LOC4346094
Genomic Map