Assay Details
| Gene Symbol: LOC4346156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793083.2 | XP_015648569.1 | 4 - 5 | 873 | 70 | |
| XM_015793084.2 | XP_015648570.1 | 5 - 6 | 768 | 70 | ||
More Information
Set Membership: |
|

LOC4346156
Genomic Map