Assay Details
Gene Symbol: LOC4346410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757188.2 | XP_015612674.1 | 2 - 3 | 447 | 74 | |
GenBank mRNA | AK061717.1 | - | 2 - 3 | 439 | 74 | |
AK065986.1 | - | 2 - 3 | 403 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
