Assay Details
Gene Symbol: LOC4346410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757188.2 | XP_015612674.1 | 6 - 7 | 735 | 72 | |
GenBank mRNA | AK061717.1 | - | 6 - 7 | 727 | 72 | |
AK065986.1 | - | 6 - 7 | 691 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
