Assay Details
Gene Symbol: LOC4346858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757149.2 | XP_015612635.1 | 8 - 9 | 2793 | 103 | |
XM_015757150.2 | XP_015612636.1 | 8 - 9 | 2794 | 103 | ||
GenBank mRNA | AK065958.1 | - | 8 - 9 | 2781 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
