Assay Details
Gene Symbol: LOC4348723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK240989.1 | - | 999 | 144 | ||
AK287809.1 | - | 1010 | 144 |
Gene Symbol: LOC4348088 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757262.2 | XP_015612748.1 | 2 - 3 | 254 | 144 | |
GenBank mRNA | AK071673.1 | - | 2 - 3 | 165 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757262.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
