Assay Details
| Gene Symbol: LOC4348782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758285.2 | XP_015613771.1 | 1 | 741 | 106 | |
| GenBank mRNA | AK073080.1 | - | 1 | 748 | 106 | |
More Information
Set Membership: |
|

LOC4348782
Genomic Map