Assay Details
| Gene Symbol: LOC9270011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759019.2 | XP_015614505.1 | 8 - 9 | 1409 | 99 | |
| GenBank mRNA | AK121234.1 | - | 1 - 2 | 62 | 99 | |
More Information
Set Membership: |
|

LOC9270011
Genomic Map