Assay Details
Gene Symbol: LOC4349420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759100.2 | XP_015614586.1 | 16 - 17 | 2187 | 133 | |
GenBank mRNA | AK103864.1 | - | 16 - 17 | 2191 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
