Assay Details
Gene Symbol: LOC4349420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759100.2 | XP_015614586.1 | 17 - 18 | 2387 | 118 | |
GenBank mRNA | AK103864.1 | - | 17 - 18 | 2391 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
