Assay Details
Gene Symbol: LOC4349420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759100.2 | XP_015614586.1 | 18 - 19 | 2554 | 145 | |
GenBank mRNA | AK103864.1 | - | 18 - 19 | 2558 | 145 | |
BT130623.1 | - | 1 - 2 | 158 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
