Assay Details
Gene Symbol: LOC4349592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761846.2 | XP_015617332.1 | 2 - 3 | 2666 | 143 | |
GenBank mRNA | AK243543.1 | - | 2 - 3 | 2627 | 143 |
Gene Symbol: LOC4351305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763902.2 | XP_015619388.1 | 2 - 3 | 2665 | 143 | |
GenBank mRNA | AK068644.1 | - | 2 - 3 | 2617 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
