Assay Details
Gene Symbol: LOC4351727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762586.2 | XP_015618072.1 | 9 - 10 | 1115 | 115 | |
GenBank mRNA | AK059540.1 | - | 2 - 3 | 97 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
