Assay Details
Gene Symbol: LOC4351727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762586.2 | XP_015618072.1 | 12 - 13 | 1456 | 83 | |
GenBank mRNA | AK059540.1 | - | 5 - 6 | 438 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
