Assay Details
| Gene Symbol: LOC4351998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762781.2 | XP_015618267.1 | 2 - 3 | 527 | 112 | |
| GenBank mRNA | AF338343.1 | - | 2 - 3 | 534 | 112 | |
| AK063847.1 | - | 2 - 3 | 487 | 112 | ||
| AK121477.1 | - | 2 - 3 | 488 | 112 | ||
More Information
Set Membership: |
|

LOC4351998
Genomic Map