Assay Details
Gene Symbol: LOC4352539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764431.2 | XP_015619917.1 | 10 - 11 | 2013 | 100 | |
GenBank mRNA | AK101383.1 | - | 10 - 11 | 2007 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
