Assay Details
Gene Symbol: LOC4352539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764431.2 | XP_015619917.1 | 12 - 13 | 3009 | 99 | |
GenBank mRNA | AB118004.1 | - | 2 - 3 | 533 | 99 | |
AK071954.1 | - | 2 - 3 | 762 | 99 | ||
AK101383.1 | - | 12 - 13 | 3003 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
