Assay Details
Gene Symbol: LOC4352539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764431.2 | XP_015619917.1 | 13 - 14 | 3371 | 61 | |
GenBank mRNA | AB118004.1 | - | 3 - 4 | 895 | 61 | |
AK071954.1 | - | 3 - 4 | 1124 | 61 | ||
AK101383.1 | - | 13 - 14 | 3365 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
