Assay Details
Gene Symbol: LOC4352765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763502.2 | XP_015618988.1 | 2 - 3 | 497 | 67 | |
GenBank mRNA | AK073789.1 | - | 2 - 3 | 396 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
