Assay Details
Gene Symbol: LOC4350166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759693.2 | XP_015615179.1 | 11 - 12 | 1310 | 69 | |
GenBank mRNA | AK099694.1 | - | 11 - 12 | 1294 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
