Assay Details
| Gene Symbol: LOC4350701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760927.2 | XP_015616413.1 | 3 - 4 | 613 | 80 | |
| XM_015760928.2 | XP_015616414.1 | 3 - 4 | 595 | 80 | ||
| GenBank mRNA | AK060533.1 | - | 2 - 3 | 235 | 80 | |
More Information
Set Membership: |
|

LOC4350701
Genomic Map