Assay Details
Gene Symbol: LOC9266255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760936.2 | XP_015616422.1 | 3 - 4 | 309 | 113 | |
XM_015760938.2 | XP_015616424.1 | 3 - 4 | 312 | 113 | ||
GenBank mRNA | AK241350.1 | - | 3 - 4 | 228 | 113 |
Gene Symbol: LOC4350701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760927.2 | XP_015616413.1 | 4 - 5 | 686 | 113 | |
XM_015760928.2 | XP_015616414.1 | 4 - 5 | 668 | 113 | ||
GenBank mRNA | AK060533.1 | - | 3 - 4 | 308 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
