Assay Details
Gene Symbol: LOC4325218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789629.2 | XP_015645115.1 | 1 - 2 | 167 | 104 | |
GenBank mRNA | AK243576.1 | - | 1 - 2 | 168 | 104 | |
BT128957.1 | - | 1 - 2 | 166 | 104 | ||
BT129829.1 | - | 1 - 2 | 168 | 104 | ||
BT131881.1 | - | 1 - 2 | 168 | 104 | ||
U43530.1 | - | 1 - 2 | 72 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015789629.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
