Assay Details
Gene Symbol: LOC4324523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765957.2 | XP_015621443.1 | 1 - 2 | 190 | 119 | |
GenBank mRNA | AK120777.1 | - | 1 - 2 | 144 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765957.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
