Assay Details
Gene Symbol: LOC4327119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791158.2 | XP_015646644.1 | 1 - 2 | 356 | 57 | |
GenBank mRNA | AK287596.1 | - | 2776 | 57 | ||
AK287669.1 | - | 1 - 2 | 291 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
