Assay Details
Gene Symbol: LOC4327119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791158.2 | XP_015646644.1 | 4 - 5 | 609 | 64 | |
GenBank mRNA | AK287596.1 | - | 3029 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
