Assay Details
Gene Symbol: LOC4325652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761569.2 | XP_015617055.1 | 10 - 11 | 1344 | 64 | |
GenBank mRNA | AK066832.1 | - | 10 - 11 | 1350 | 64 | |
AK068959.1 | - | 10 - 11 | 1300 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
