Assay Details
Gene Symbol: LOC4325652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761569.2 | XP_015617055.1 | 13 - 14 | 1747 | 127 | |
GenBank mRNA | AB117991.1 | - | 3 - 4 | 295 | 127 | |
AK066832.1 | - | 13 - 14 | 1753 | 127 | ||
AK068959.1 | - | 13 - 14 | 1703 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
