Assay Details
Gene Symbol: LOC4325652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761569.2 | XP_015617055.1 | 14 - 15 | 1916 | 87 | |
GenBank mRNA | AB117991.1 | - | 4 - 5 | 464 | 87 | |
AK066832.1 | - | 14 - 15 | 1922 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
