Assay Details
Gene Symbol: LOC4325652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761569.2 | XP_015617055.1 | 15 - 16 | 1990 | 80 | |
GenBank mRNA | AB117991.1 | - | 5 - 6 | 538 | 80 | |
AK066832.1 | - | 15 - 16 | 1996 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
