Assay Details
Gene Symbol: LOC4325652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761569.2 | XP_015617055.1 | 3 - 4 | 536 | 119 | |
GenBank mRNA | AK068959.1 | - | 3 - 4 | 492 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
