Assay Details
Gene Symbol: LOC4325652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761569.2 | XP_015617055.1 | 9 - 10 | 1145 | 128 | |
GenBank mRNA | AK066832.1 | - | 9 - 10 | 1151 | 128 | |
AK068959.1 | - | 9 - 10 | 1101 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
