Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 11 - 12 | 1346 | 71 | |
| GenBank mRNA | AK060358.1 | - | 4 - 5 | 289 | 71 | |
| AK121770.1 | - | 11 - 12 | 1256 | 71 | ||
| KC140122.1 | - | 11 - 12 | 1221 | 71 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map