Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 17 - 18 | 1794 | 74 | |
| GenBank mRNA | AK060358.1 | - | 10 - 11 | 737 | 74 | |
| AK072842.1 | - | 17 - 18 | 1702 | 74 | ||
| AK121770.1 | - | 17 - 18 | 1704 | 74 | ||
| KC140122.1 | - | 17 - 18 | 1669 | 74 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map