Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 6 - 7 | 931 | 90 | |
| GenBank mRNA | AK072842.1 | - | 6 - 7 | 841 | 90 | |
| AK121770.1 | - | 6 - 7 | 841 | 90 | ||
| KC140122.1 | - | 6 - 7 | 806 | 90 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map