Assay Details
Gene Symbol: LOC4324278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757095.1 | XP_015612581.1 | 8 - 9 | 1523 | 82 | |
GenBank mRNA | AK120694.1 | - | 6 - 7 | 2767 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
